Anti-Emmprin Polyclonal Antibody
Leverandør: Boster Bio
PB9247
BSBTPB9247EA 6475 NOK
BSBTPB9247
Anti-Emmprin Polyclonal Antibody
Antistoffer
Polyclonal antibody for 575 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: Flow Cytometry. Reactive species: Human. 575 information: Molecular Weight: 42200 MW; Subcellular Localization: Cell membrane ; Single-pass type I membrane protein . Melanosome . Colocalizes with SLC16A1 and SLC16A8. Identified by mass spectrometry in melanosome fractions from stage I to stage IV; Tissue Specificity: Present only in vascular endothelium in non- neoplastic regions of the brain, whereas it is present in tumor cells but not in proliferating blood vessels in malignant gliomas.