Anti-ACO2 Rabbit Polyclonal Antibody
Leverandør: Boster Bio
PB9973
BSBTPB9973EA 6475 NOK
BSBTPB9973
Anti-ACO2 Rabbit Polyclonal Antibody
Antistoffer
Aconitase 2, mitochondrial is a protein that in humans is encoded by the ACO2 gene. The protein encoded by this gene belongs to the aconitase/IPM isomerase family. It is an enzyme that catalyzes the interconversion of citrate to isocitrate via cis-aconitate in the second step of the TCA cycle. This protein is encoded in the nucleus and functions in the mitochondrion. It was found to be one of the mitochondrial matrix proteins that are preferentially degraded by the serine protease 15(PRSS15), also known as Lon protease, after oxidative modification.
1. "Entrez Gene: Aconitase 2,More Product Information