Anti-Peptide YY Chicken Polyclonal Antibody
Leverandør: Biosensis
C-1537-100
BSENC-1537-100EA 3925 NOK
BSENC-1537-100
Anti-Peptide YY Chicken Polyclonal Antibody
Antistoffer
Peptide YY (PYY) is secreted from endocrine cells in the lower small intestine, colon and pancreas. PYY inhibits exocrine pancreatic secretion, has a vasoconstrictory action and inhibitis jejunal and colonic mobility (Ref: SWISS-Prot).
Antibody recognizes Peptide YY in ELISAs, other applications have not been tested. This format is biotinylated and can be used as a detector antibody in ELISAs when paired with C-1538
Application Information:
ELISA . Suggested dilution at 1:2,000 to 1:10,000 (detection antibody). Biosensis recommends that the optimal working dilution should be determined byMore Product Information