Anti-Peptide YY Chicken Polyclonal Antibody

Leverandør: Biosensis
C-1538-100
BSENC-1538-100EA 3775 NOK
BSENC-1538-100
Anti-Peptide YY Chicken Polyclonal Antibody
Antistoffer
Peptide YY (PYY) is secreted from endocrine cells in the lower small intestine, colon and pancreas. PYY inhibits exocrine pancreatic secretion, has a vasoconstrictory action and inhibitis jejunal and colonic mobility (Ref: SWISS-Prot).

Antibody recognizes Peptide YY in ELISAs, other applications have not been tested.
Application Information:
ELISA . Suggested dilution at 1:200 to 1:1,000 (coating antibody) and 1:2,000 to 1:5,000 (detection antibody) .

Type: Primary
Antigen: Peptide YY
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminus of Human mature Peptide YY (3-18More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR