Anti-IgA Goat Polyclonal Antibody (CF™ 633)
Leverandør: Biotium
20427-50UL 20427-500UL
BTIU20427-50ULEA 1500 NOK
BTIU20427-50UL BTIU20427-500UL
Anti-IgA Goat Polyclonal Antibody (CF™ 633)
Antistoffer
Goat anti-human IgA reacts specifically with IgA heavy chains (α chains) and not with immunoglobulin light chains. To minimize crossreactivity, it has been adsorbed human IgG prior to conjugation. This conjugate is labeled with CF™633 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Far-red fluorescent CF™633 (Ex/Em 630/650 nm) produces brighter antibody conjugates than Alexa Fluor® 647, and has unmatched photostability.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GEMore Product Information