Anti-VIM Mouse Monoclonal Antibody (Purified, BSA-free) [clone: LN-6]
Leverandør: Biotium
BNUM0060-50
BTIUBNUM0060-50EA 5125 NOK
BTIUBNUM0060-50
Anti-VIM Mouse Monoclonal Antibody (Purified, BSA-free) [clone: LN-6]
Antistoffer
This MAb reacts with a 58 kDa protein identified as vimentin. It reacts with a non-hematopoietic epitope of vimentin and shows no cross-reaction with other closely related intermediate filament proteins (IFP's) such as desmin, keratin, neurofilament, and glial fibrillary acid protein.Vimentin is ubiquitously expressed in mesenchymal cells such as fibroblasts, smooth muscle cells, and endothelium. Antibody against vimentin is useful as part of an antibody panel for differential diagnosis of tumors of unknown origin.Ab-2 does not react with leukocyte common antigen-positive tissues such asMore Product Information