Anti-ESR1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ER505]

Leverandør: Biotium
BNUM0505-50
BTIUBNUM0505-50EA 5125 NOK
BTIUBNUM0505-50
Anti-ESR1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ER505]
Antistoffer
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. This MAb is excellent for staining of formalin-fixed, paraffin-embedded breast carcinomas.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: ER505
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR