Anti-HSPB1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: HSPB1/774]

Leverandør: Biotium
BNUM0774-50
BTIUBNUM0774-50EA 5125 NOK
BTIUBNUM0774-50
Anti-HSPB1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: HSPB1/774]
Antistoffer
This antibody recognizes a 24-27 kDa estrogen-regulated protein, identified as heat shock protein 27 (hsp27). Hsp27 was recently found to be identical to the estrogen-induced p29 and 24K protein. About 50% of breast carcinomas are positive for hsp27 especially those that are also positive for estrogen and/or progesterone receptor. HSP27 has also been implicated in drug resistance in cancer cells.

Type: Primary
Antigen: HSPB1
Clonality: Monoclonal
Clone: HSPB1/774
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken, Chimpanzee, Human, Mouse, Rat, Sheep
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR