Anti-TYRP1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: TA99]

Leverandør: Biotium
BNUM0806-50
BTIUBNUM0806-50EA 5125 NOK
BTIUBNUM0806-50
Anti-TYRP1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: TA99]
Antistoffer
This antibody reacts with a 75 kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TRP-1). It is involved in melanin synthesis. TRP1 is present on the melanosomal membranes of melanoma, normal melanocytes and nevi.Recent evidence suggests that TRP-1 is involved in maintaining stability of tyrosinase protein and modulating its catalytic activity. TRP-1 is also involved in maintenance of melanosome ultrastructure and affects melanocyte proliferation and cell death.

Type: Primary
Antigen: TYRP1
Clonality: Monoclonal
Clone: TA99
Conjugation: Purified, BSA-free
Epitope:More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR