Anti-TGFA Mouse Monoclonal Antibody (Purified, BSA-free) [clone: TGFA/1119]

Leverandør: Biotium
BNUM1119-50
BTIUBNUM1119-50EA 5125 NOK
BTIUBNUM1119-50
Anti-TGFA Mouse Monoclonal Antibody (Purified, BSA-free) [clone: TGFA/1119]
Antistoffer
This antibody reacts with the C-terminus of TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. TGF alpha (aa50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.

Type: Primary
Antigen: TGFA
Clonality: Monoclonal
Clone: TGFA/1119
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR