Anti-UGT1A9 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: UGT1A9/1229]

Leverandør: Biotium
BNUM1229-50
BTIUBNUM1229-50EA 5125 NOK
BTIUBNUM1229-50
Anti-UGT1A9 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: UGT1A9/1229]
Antistoffer
This antibody recognizes a protein of about 60 kDa, which is identified as human UGT1A9. It does not cross-react with the other UGT1A isoforms including UGT1A7, UGT1A8, and UGT1A10 and shows a high degree of specificity. UGT1A9 is a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This MAb binds to human hepatocytes and the majority of human hepatocellular carcinomas (HCC s). In frozen sections, it stains hepatic cells and may be usedMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol UGT1A9
Antigen-navn UDP glucuronosyltransferase 1 family, polypeptide A9
Environmentally Preferable
Antigen synonymer UDPGT 1-9|UGT1-09|Ugt1a9|UDP-glucuronosyltransferase 1A9 (UGT1A9)|UGT-1I|GNT1|UDP-glucuronosyltransferase 1-I|lugP4|UGT1.9|HLUGP4|UGT1*9|UGT1I|UDPGT
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon Clone UGT1A9/1229
Reaktivitet Human
Vert Mouse
Gen-ID 554822
Isotype IgG2b kappa
Rensing BSA-free
Konsentrasjon 1 mg/mL
Lagringsbuffer PBS/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR