LabFlame+ Bunsen Burner, with Integrated NIMI (AA)
Leverandør: BioTool
LF8.204.000
BTOLLF8.204.000EA 11300 NOK
BTOLLF8.204.000
LabFlame+ Bunsen Burner, with Integrated NIMI (AA)
Termostater Gassbrennere
The LabFlame+  is optimised for use in microbiology laboratories. Its stainless steel housing is made of one piece, free of any gaps or rims and therefore easy to clean and fully UV resistant. Its optimised air-flow system allows a uniform and constant flame. Connectable to any kind of gas sources. Ideal for use in cleanroom workbenches and laboratories.
  • The animated and high-contrast graphic display can be easily read under all lighting conditions due to the blue background illumination
  • The self-explanatory, language-less symbol menu 