Petri dish platinum electrode
Leverandør: BTX
45-0507 45-0505
BTXI45-0507EA 27300 NOK
BTXI45-0507 BTXI45-0505
Petri dish platinum electrode
Elektroder Elektroporasjonselektroder
This tissue chamber is specifically designed to handle ex-vivo tissue samples that are either larger than normal or have a unique shape making it difficult to transfect using other standard electrodes. This is an efficient method to deliver genes and drugs to a wide range of tissue types including cornea, muscle and skin.
  • Chambers are available in two widths (15 mm and 5 mm) to accommodate many tissue sample sizes
  • The reusable chamber of a lab grade Pyrex glass petri dish and two platinum electrodes embedded in an inert silicone
  • Rectangular chamber that