Petri dish platinum electrodes
Leverandør: BTX
45-2059 45-2060
BTXI45-2059EA 32825 NOK
BTXI45-2059 BTXI45-2060
Petri dish platinum electrodes
Elektroder Elektroporasjonselektroder
These reusable oocyte Petri electrodes are ideal for electroporation of oocytes, embryos, or other cell types in small volumes.
  • Electrodes are permanently attached to a glass Petri dish and come with a glass lid. Designed to easily fit on an inverted microscope stage to allow easy observation of cells during electroporation.