Barrel valve
Leverandør: BURKLE
0536-0010
BURK0536-0010EA 3750 NOK
BURK0536-0010
Barrel valve
Haner
The barrel valve for opening the barrel closure as well as emptying and reclosing it even with lying, full barrels without having to move them. Also used for highly viscous liquids.