Kegs, wide neck, UN approved, with screw lid
Leverandør: BURKLE
1105-0026 1105-0003 1105-0006 1105-0010 1105-0033
BURK1105-0026EA 2225 NOK
BURK1105-0026 216-5385 216-4596 BURK1105-0010 BURK1105-0033
Kegs, wide neck, UN approved, with screw lid
Tønner, fat, trommer
HDPE white containers with red screw lid.
  • Temperature resistant from −20 to + 80°C
  • Cap has integrated rubber gasket to ensure an airtight and watertight seal
  • UN approved
The wide-necked container is approved for bulk goods. Used for disposal or storage. For liquids, powders or granules. Can be closed tightly and sealed.