Packaging Bags with Closure
Leverandør: BURKLE
2348-0006 2348-0008 2348-0010 2348-0003 2348-0001 2348-0004 2348-0007 2348-0002 2348-0009
113-5411EA 510 NOK
113-5411 113-5412 113-5413 113-5415 BURK2348-0001 BURK2348-0004 BURK2348-0007 BURK2348-0002 BURK2348-0009
Packaging Bags with Closure
Poser Oppbevaringsposer
Bags with the patented zipper are for ideal for storing, collecting, protecting and shipping of specimens, samples and sales products.
  • Made of LDPE
  • Film has good tear resistance
  • Crystal-clear
  • Waterproof
Contents remain dust-free.