Tanks, cylindrical, with lid, LaboPlast®
Leverandør: BURKLE
3490-0144 3490-1606 3490-2506 3490-0855 3490-1005 3490-0305 3490-1206 3490-0355 3490-0805 3490-2006 3490-0505 3490-1106 3490-3006 3490-8507
BURK3490-0144EA 4275 NOK
BURK3490-0144 BURK3490-1606 BURK3490-2506 BURK3490-0855 BURK3490-1005 BURK3490-0305 BURK3490-1206 BURK3490-0355 BURK3490-0805 BURK3490-2006 BURK3490-0505 BURK3490-1106 BURK3490-3006 BURK3490-8507
Tanks, cylindrical, with lid, LaboPlast®
Store beholdere Store beholdere, sylindriske
Containers in LDPE, transparent, with lids, for various applications. Produced seamlessly and stress relieved with the LaboPlast® sintering method.
  • Round shaped
  • Coloured black for UV protection
  • Rupture-proof
The wall thickness increases downwardly. This guarantess optimum ratio of wall thickness to filling load.
Bestillingsinformasjon: Lids have to be ordered separatly (see accessories).