Prøvetakere, MicroSampler
Leverandør: BURKLE
5307-2055 5307-0205 5307-2085 5307-0210 5307-1055 5307-1085 5307-1120 5307-0102 5307-0105 5307-0110 5307-2120 5307-0202 5307-0101
331-2191EA 10175 NOK
331-2191 331-2192 331-0030 331-0026 331-0027 331-0028 331-0029 331-0022 331-0023 331-0024 331-0031 331-0025 BURK5307-0101
Prøvetakere, MicroSampler
Prøvetakere Prøvetakere til faststoff
Stainless steel AISI 316 L (1,4404). The MicroSampler has been specially designed for the sampling of small quantities of powder.
  • Point sampling
  • Various lengths and volumes are available
  • Tip can be removed for easy cleaning
  • Additional tips are available in different sizes
  • Sterilisable
Volumes of 0,2 ml, 0,5 ml, 1,0 ml, 2,0 ml, 5,0 ml, and 10 ml can be sampled easily, quickly, and cleanly from different sampling depths from sacks or containers.
The tips of the MicroSampler are interchangeable. This means that tips with different volumes can be used.