All-layer drill
Leverandør: BURKLE
5310-0150
BURK5310-0150EA 43525 NOK
BURK5310-0150
All-layer drill
Prøvetakere Prøvetakere til faststoff
This sampler with drilling blades makes it easy to take samples of media, even those with high density, and every type of highly compacted powders and granulates.
This special zone collector is suitable for heavy and compact substances – in large containers, mixed containers, big bags in the building materials, chemical, pharmaceutical, food and luxury items industries.