Sample spoons
Leverandør: BURKLE
5320-0150 5320-0180 5320-0210 5320-0300 5320-0400 5320-0500
300-0019EA 185 NOK
300-0019 BURK5320-0180 300-0020 BURK5320-0300 300-0021 BURK5320-0500
Sample spoons
Skjeer
Made of AISI 304 (1.4301) stainless steel. This specially shaped spoon reaches almost every corner in containers and prevents samples from trickling out.
  • For withdrawing small sample quantities
  • Largely chemical resistant (not against acids and alkalis)