Measuring spoons, stainless steel
Leverandør: BURKLE
5334-8000
BURK5334-8000EA 460 NOK
BURK5334-8000
Measuring spoons, stainless steel
Skjeer
Stainless steel V2A. Set of four measuring spoons. Ideal for fast and easy measuring from 1,25 to 15 ml.
  • High-gloss finished, handle matt polished
  • Volume is engraved on the handle of each measuring spoon