Tamper-proof sample bags, SteriBag Premium
Leverandør: BURKLE
5344-8000
BURK5344-8000EA 5475 NOK
BURK5344-8000
Tamper-proof sample bags, SteriBag Premium
Poser Prøveposer
The SteriBag Premium sample bags uniquely combine a high degree of security, sterility, tamper evident closure and traceability, they are used to securely transport and for the storage of samples.
  • Sterilised by gamma rays (SAL 10⁻⁶)
  • With integrated tamper-proof closure for manipulation evidence
  • Unique identification via tracking number
  • Suitable for the use with foodstuffs
After sampling, the sample can be filled directly into the sterile sample bag. The tamper-evident closure seals the bag liquid-tight and secure. When trying to open the closure again,More Product Information