Powder samplers, PharmaPicker
Leverandør: BURKLE
5346-1000 5346-0010 5346-2000
BURK5346-1000EA 13225 NOK
BURK5346-1000 312-0102 BURK5346-2000
Powder samplers, PharmaPicker
Prøvetakere Prøvetakere til faststoff
PharmaPicker is designed for sampling very small quantities of highly potent or active powders for analysis. The smallest sample quantities can be collected with the PharmaPicker.
  • Available separately or in a handy transport box
  • Electro-polished stainless steel
  • Can be extended repeatedly
Leveringsinformasjon: PharmaPicker case includes collection cylinders in both sizes, extension rods of 50 and 100 cm, a handle, volume tips (alternative sample volumes on request), and a transport box.