Målnedsenkingssylinder
Leverandør: BURKLE
5365-6000
BURK5365-6000EA 14525 NOK
BURK5365-6000
Målnedsenkingssylinder
Prøvetakere Væskeprøvetakere
Precise liquid samples can be taken with this immersion cylinder. When the target has been reached, open the cover and pour out the sample.
  • Screw-off top enables easy cleaning
  • Air valve at the end of the tube
  • Venting lowering cable and cable reel