Micro spatula, stainless steel
Leverandør: BURKLE
5386-0001
BURK5386-0001EA 130 NOK
BURK5386-0001
Micro spatula, stainless steel
Spatler
The micro spatula can be used for reliably measuring, sampling, or processing small amounts of powder, chemicals, pastes, creams, or active ingredients.
  • Stainless steel AISI 304 (1.4301)
  • High-gloss polished
The laboratory scoop is manufactured entirely without edges, crevices, or undercuts. This eliminates the possibility of impurities or cross-contamination, as no product residue or cleaning agents can be deposited. The micro spatula is ideal for use in the pharmaceutical industry as well as food and beverage, and cosmetics.