Sample container, with screw cap
Leverandør: BURKLE
6205-0180
BURK6205-0180EA 3125 NOK
BURK6205-0180
Sample container, with screw cap
Prøvecontainere Beholdere, generelt bruk
PP or PFA, translucent.
  • Excellent chemical resistance
  • Thick sides
  • Sample container PFA not suitable for food
Sample box with screw lid. Ideal for taking samples and transport. Available in two material qualities: highly transparent PP for powder, pastes, liquids, or PFA for super pure collection of samples. Emptying without residues because there are no crevices.