Bokser, vid åpning, runde, med skrukork
Leverandør: BURKLE
6282-2000 6282-1000 6282-0500 6282-0100 6282-0250 6282-0750 6282-1500
BURK6282-2000EA 210 NOK
BURK6282-2000 BURK6282-1000 BURK6282-0500 BURK6282-0100 BURK6282-0250 BURK6282-0750 BURK6282-1500
Bokser, vid åpning, runde, med skrukork
Krukker Bokser, generell bruk
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Screw lid made of PP with seal made of PE
Wide opening makes it ideally suited for storing or shipping powders and granules.