Sedimentation cones, Imhoff type
Leverandør: BURKLE
7118-1000
BURK7118-1000EA 1275 NOK
BURK7118-1000
Sedimentation cones, Imhoff type
Trakter Immhoff-trakter
Styrene-acrylonitrile (SAN), PP screw plug.
  • As transparent as glass
  • Easy draining via screw plug
Graduation: 0 to 2 ml:0,1/ 2 to 10 ml:0,5/ 10 to 40 ml:1/ 40 to 100 ml:2/ 100 to 1000 ml:10