Skuffe i én del for næringsmidler
Leverandør: BURKLE
8300-0011
BURK8300-0011EA 3275 NOK
BURK8300-0011
Skuffe i én del for næringsmidler
Spader
PP, white. This 1-part shovel is ideal for use in the food sector.
  • PP is easy to clean, smooth and pore-free
  • Can also be used for boiling products
  • Autoclavable at 121 °C
  • FDA approved