Skuffe i to deler for næringsmidler
Leverandør: BURKLE
8300-2010
BURK8300-2010EA 1975 NOK
BURK8300-2010
Skuffe i to deler for næringsmidler
Spader
PP, white. This 2-part shovel is ideal for use in the food sector.
  • PP is easy to clean, smooth and pore-free
  • Can also be used for boiling products
  • Autoclavable at 121 °C
  • FDA approved