Anti-CLDN4 Rabbit Polyclonal Antibody [clone: P192]

Leverandør: Bioworld Technology
BS1068
BWRLBS1068EA 3375 NOK
BWRLBS1068
Anti-CLDN4 Rabbit Polyclonal Antibody [clone: P192]
Antistoffer
Synthetic peptide, corresponding to amino acids 171-220 of Human Claudin-4.

Claudin 4 (P192) pAb detects endogenous levels of Claudin 4 protein.

Type: Primary
Antigen: CLDN4
Clonality: Polyclonal
Clone: P192
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR