Anti-OPN Rabbit Polyclonal Antibody [clone: L285]

Leverandør: Bioworld Technology
BS1264
BWRLBS1264EA 3375 NOK
BWRLBS1264
Anti-OPN Rabbit Polyclonal Antibody [clone: L285]
Antistoffer
Synthetic peptide, corresponding to amino acids 251-300 of Human OPN.

Osteopontin/SPP1 (L285) pAb detects endogenous levels of Osteopontin/SPP1 protein.

Type: Primary
Antigen: OPN
Clonality: Polyclonal
Clone: L285
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR