Anti-UCHL1 Rabbit Polyclonal Antibody [clone: R202]

Leverandør: Bioworld Technology
BS1293
BWRLBS1293EA 3375 NOK
BWRLBS1293
Anti-UCHL1 Rabbit Polyclonal Antibody [clone: R202]
Antistoffer
Synthetic peptide, corresponding to amino acids 171-220 of Human UCHL1.

PGP9.5 (R202) pAb detects endogenous levels of PGP9.5 protein.

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone: R202
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR