Anti-RPA2 Rabbit Polyclonal Antibody [clone: A17]

Leverandør: Bioworld Technology
BS1692
BWRLBS1692EA 3375 NOK
BWRLBS1692
Anti-RPA2 Rabbit Polyclonal Antibody [clone: A17]
Antistoffer
Synthetic peptide, corresponding to the N-terminus of Human RPA2.

RPA2 (A17) pAb detects endogenous levels of RPA2 protein.

Type: Primary
Antigen: RPA2
Clonality: Polyclonal
Clone: A17
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR