Anti-GRK1 Rabbit Polyclonal Antibody [clone: A17]

Leverandør: Bioworld Technology
BS3251
BWRLBS3251EA 3375 NOK
BWRLBS3251
Anti-GRK1 Rabbit Polyclonal Antibody [clone: A17]
Antistoffer
Synthetic peptide, corresponding to the N-terminal of Human GRK1.

GRK1 (A17) pAb detects endogenous levels of GRK1 protein.

Type: Primary
Antigen: GRK1
Clonality: Polyclonal
Clone: A17
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR