Anti-p-Rb Rabbit Polyclonal Antibody [clone: S795]

Leverandør: Bioworld Technology
BS4165
BWRLBS4165EA 3675 NOK
BWRLBS4165
Anti-p-Rb Rabbit Polyclonal Antibody [clone: S795]
Antistoffer
Synthetic phosphopeptide derived from human Rb around the phosphorylation site of Serine 795.

p-Rb (S795) pAb detects endogenous levels of Rb protein only when phosphorylated at Ser795

Type: Primary
Antigen: p-Rb
Clonality: Polyclonal
Clone: S795
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol p-Rb
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Klon S795
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype
Western blot Yes
ImmunoKjemi Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR