Anti-p-PDLIM5 Rabbit Polyclonal Antibody [clone: Y251]

Leverandør: Bioworld Technology
BS4820
BWRLBS4820EA 3675 NOK
BWRLBS4820
Anti-p-PDLIM5 Rabbit Polyclonal Antibody [clone: Y251]
Antistoffer
Synthetic phosphopeptide derived from human PDLIM5 around the phosphorylation site of Tyrosine 251.

p-PDLIM5 (Y251) pAb detects endogenous levels of PDLIM5 protein only when phosphorylated at Tyr251.

Type: Primary
Antigen: p-PDLIM5
Clonality: Polyclonal
Clone: Y251
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR