Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: Y15]

Leverandør: Bioworld Technology
BS5058
BWRLBS5058EA 3675 NOK
BWRLBS5058
Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: Y15]
Antistoffer
Synthetic phosphopeptide derived from human Cdk1/Cdc2 around the phosphorylation site of Serine 795.

p-Cdk1/Cdc2 (Y15) pAb detects endogenous levels of Cdk1/Cdc2 protein only when phosphorylated at Tyr15.

Type: Primary
Antigen: p-Cdk1/Cdc2
Clonality: Polyclonal
Clone: Y15
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol p-Cdk1/Cdc2
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Klon Y15
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype
Western blot Yes
ImmunoKjemi Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR