Anti-GPR105 Rabbit Polyclonal Antibody

Leverandør: Bioworld Technology
BS5738
BWRLBS5738EA 3375 NOK
BWRLBS5738
Anti-GPR105 Rabbit Polyclonal Antibody
Antistoffer
Synthetic peptide, corresponding to amino acids 197-242 of Human GPR105.

GPR105 pAb detects endogenous levels of GPR105 protein.

Type: Primary
Antigen: GPR105
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol GPR105
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype
Western blot Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR