Anti-KIF4A Rabbit Polyclonal Antibody

Leverandør: Bioworld Technology
BS5774
BWRLBS5774EA 3375 NOK
BWRLBS5774
Anti-KIF4A Rabbit Polyclonal Antibody
Antistoffer
Synthetic peptide, corresponding to amino acids 1187-1232 of Human KIF4A.

KIF4A pAb detects endogenous levels of KIF4A protein.

Type: Primary
Antigen: KIF4A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR