Anti-MPP10 Rabbit Polyclonal Antibody

Leverandør: Bioworld Technology
BS5799
BWRLBS5799EA 3375 NOK
BWRLBS5799
Anti-MPP10 Rabbit Polyclonal Antibody
Antistoffer
Synthetic peptide, corresponding to amino acids 126-171 of Human MPP10.

MPP10 pAb detects endogenous levels of MPP10 protein.

Type: Primary
Antigen: MPP10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol MPP10
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype
Western blot Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR