Anti-Actived-CASP3 p17 Rabbit Polyclonal Antibody

Leverandør: Bioworld Technology
BS7004
BWRLBS7004EA 3675 NOK
BWRLBS7004
Anti-Actived-CASP3 p17 Rabbit Polyclonal Antibody
Antistoffer
Synthetic peptide conjugated to KLH derived from within residues 150 - 200 of Human active Caspase 3.

Actived-Caspase-3 p17 pAb detects endogenous levels of Actived-Caspase-3 p17 protein.

Type: Primary
Antigen: Actived-CASP3 p17
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestill nå

SPESIFIKASJONER

Antigensymbol Actived-CASP3 p17
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Reaktivitet Human,Rat,Mouse
Vert Rabbit
Isotype
Western blot Yes
ImmunoKjemi Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR