Anti-CASP2 p18 Rabbit Polyclonal Antibody [clone: T325]

Leverandør: Bioworld Technology
BS7072
BWRLBS7072EA 3375 NOK
BWRLBS7072
Anti-CASP2 p18 Rabbit Polyclonal Antibody [clone: T325]
Antistoffer
Synthetic peptide conjugated to KLH derived from within residues 300 - 350 of Human active Caspase 2.

Caspase-2 p18 (T325) pAb detects endogenous levels of Caspase-2 p18 protein.

Type: Primary
Antigen: CASP2 p18
Clonality: Polyclonal
Clone: T325
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR