Anti-CASP3 p17 Rabbit Polyclonal Antibody [clone: D175]

Leverandør: Bioworld Technology
BS7073
BWRLBS7073EA 3375 NOK
BWRLBS7073
Anti-CASP3 p17 Rabbit Polyclonal Antibody [clone: D175]
Antistoffer
Synthetic peptide conjugated to KLH derived from within residues 150 - 200 of Human active Caspase 3.

Caspase-3 p17 (D175) pAb detects endogenous levels of the large fragment (17 kDa) of activated caspase-3 resulting from cleavage adjacent to Asp175. This antibody does not recognize full length caspase-3 or other cleaved caspases.

Type: Primary
Antigen: CASP3 p17
Clonality: Polyclonal
Clone: D175
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR