Anti-Histone H3 Rabbit Polyclonal Antibody [clone: Mono-Methyl K27]

Leverandør: Bioworld Technology
BS7235
BWRLBS7235EA 4900 NOK
BWRLBS7235
Anti-Histone H3 Rabbit Polyclonal Antibody [clone: Mono-Methyl K27]
Antistoffer
Synthetic peptide conjugated to KLH derived from within residues 1-100 of Human Histone H3, mono methylated at K27.

Histone H3 (Mono-Methyl K27) pAb detects endogenous levels of histone H3 only when mono-methylated at Lys27.

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone: Mono-Methyl K27
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human; Mouse; Rat; Other (Wide Range)
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR