Anti-Histone H3 Rabbit Polyclonal Antibody [clone: sDi-Methyl R2]

Leverandør: Bioworld Technology
BS7243
BWRLBS7243EA 4900 NOK
BWRLBS7243
Anti-Histone H3 Rabbit Polyclonal Antibody [clone: sDi-Methyl R2]
Antistoffer
Synthetic peptide conjugated to KLH derived from within residues 1-100 of Human Histone H3, sDi-methylated at R2.

Histone H3 (sDi-Methyl R2) pAb detects endogenous levels of histone H3 only when symmetric Di-Methyl at Arg2.

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone: sDi-Methyl R2
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human; Mouse; Rat; Other (Wide Range)
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR