Anti-Histone H3 Rabbit Polyclonal Antibody [clone: aDi-Methyl R17]

Leverandør: Bioworld Technology
BS7247
BWRLBS7247EA 4900 NOK
BWRLBS7247
Anti-Histone H3 Rabbit Polyclonal Antibody [clone: aDi-Methyl R17]
Antistoffer
Synthetic peptide conjugated to KLH derived from within residues 1-100 of Human Histone H3, aDi-methylated at R17.

Histone H3 (aDi-Methyl R17) pAb detects endogenous levels of histone H3 only when asymmetric Di-Methyl at Arg17.

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone: aDi-Methyl R17
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human; Mouse; Rat; Other (Wide Range)
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR