Anti-Histone H3 Rabbit Polyclonal Antibody [clone: aDi-Methyl R2]

Leverandør: Bioworld Technology
BS7511
BWRLBS7511EA 4900 NOK
BWRLBS7511
Anti-Histone H3 Rabbit Polyclonal Antibody [clone: aDi-Methyl R2]
Antistoffer
Synthetic peptide conjugated to KLH derived from within residues 1-100 of Human Histone H3, aDi-methylated at R2.

Histone H3 (aDi-Methyl R17) pAb detects endogenous levels of histone H3 only when asymmetric Di-Methyl at Arg2.

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone: aDi-Methyl R2
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat; Other (Wide Range)
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR