Anti-Histone H3 Rabbit Polyclonal Antibody [clone: Di-Methyl K14 ]

Leverandør: Bioworld Technology
BS7524
BWRLBS7524EA 4900 NOK
BWRLBS7524
Anti-Histone H3 Rabbit Polyclonal Antibody [clone: Di-Methyl K14 ]
Antistoffer
Synthetic peptide conjugated to KLH derived from within residues 1-100 of Human Histone H3, di methylated at K14.

Histone H3 (Di-Methyl K14) pAb detects endogenous levels of histone H3 only when Di-methylated at Lys14.

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone: Di-Methyl K14
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat; Other (Wide Range)
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR